CH4636885264 ChemBridge Corporation

4-methyl-2-morpholinecarboxylic acid hydrochloride

  • MDL number MFCD06738959
  • Cas Number : 841274-05-3
  • Molecular Weight : 181.6174

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4078099

Safety Information

MSDS https://www.hit2lead.com/sds/4078099_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart