CH4721366958 ChemBridge Corporation

2,5,6-trimethyl-3-pyridinamine dihydrochloride

  • MDL number MFCD30718769
  • Cas Number : 2089257-49-6
  • Molecular Weight : 209.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin RU
Product Link https://www.hit2lead.com/building-blocks/4086097

Safety Information

MSDS https://www.hit2lead.com/sds/4086097_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart