CH4721366962 ChemBridge Corporation

7-bromo-1-benzothiophene

  • MDL number MFCD06657729
  • Cas Number : 1423-61-6
  • Molecular Weight : 213.0943

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4029546

Safety Information

MSDS https://www.hit2lead.com/sds/4029546_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart