CH4721367016 ChemBridge Corporation

(2-bromo-3-fluorophenyl)acetic acid

  • MDL number MFCD09864672
  • Cas Number : 958454-33-6
  • Molecular Weight : 233.0344

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4108254

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4108254
Availability details for
Quantity
  • Availability
  • Add to Cart