CH4721367029 ChemBridge Corporation

[4-(difluoromethoxy)-3-methylphenyl]boronic acid

  • MDL number MFCD11036079
  • Cas Number : 958451-72-4
  • Molecular Weight : 201.97

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin RU
Product Link https://www.hit2lead.com/building-blocks/4108269

Safety Information

MSDS https://www.hit2lead.com/sds/4108269_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart