CH4979588637 ChemBridge Corporation

6-azaspiro[3.4]octan-5-one

  • MDL number MFCD19219361
  • Cas Number : 1193-30-2
  • Molecular Weight : 125.1683

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4035729

Safety Information

MSDS https://www.hit2lead.com/sds/4035729_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart