CH4979588725 ChemBridge Corporation

cis-3-aminocyclohexanol

  • MDL number MFCD09839763
  • Cas Number : 6982-42-9
  • Molecular Weight : 115.1735

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4108327

Safety Information

MSDS https://www.hit2lead.com/sds/4108327_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart