CH4979588729 ChemBridge Corporation

{[5-(ethylthio)-1,3,4-oxadiazol-2-yl]methyl}amine hydrochloride

  • MDL number MFCD00770573
  • Cas Number : 1094417-07-8
  • Molecular Weight : 195.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Salt Data HCl
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5147135

Safety Information

MSDS https://www.hit2lead.com/sds/5147135_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart