CH4979588733 ChemBridge Corporation

N-ethyl-2-adamantanamine hydrochloride

  • MDL number MFCD09971526
  • Cas Number : 10523-70-3
  • Molecular Weight : 215.7628

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6044024

Safety Information

MSDS https://www.hit2lead.com/sds/6044024_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart