CH4979588735 ChemBridge Corporation

methyl 1-ethyl-4-oxo-3-piperidinecarboxylate hydrochloride

  • MDL number MFCD09836251
  • Cas Number : 24346-37-0
  • Molecular Weight : 221.6812

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6657556

Safety Information

MSDS https://www.hit2lead.com/sds/6657556_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart