CH4979588742 ChemBridge Corporation

(4-fluorobenzyl)(4-pyridinylmethyl)amine hydrochloride

  • MDL number MFCD06002298
  • Cas Number : 1052418-35-5
  • Molecular Weight : 252.7151

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/7934997

Safety Information

MSDS https://www.hit2lead.com/sds/7934997_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart