CH4979588749 ChemBridge Corporation

4-{[(2-furylmethyl)amino]methyl}benzoic acid hydrochloride

  • MDL number MFCD08068143
  • Cas Number : 1050591-26-8
  • Molecular Weight : 267.7082

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9012014

Safety Information

MSDS https://www.hit2lead.com/sds/9012014_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart