CH4H03CD9AC5 ChemBridge Corporation

[2-(2-pyrazinyl)ethyl]amine hydrochloride hydrate

  • MDL number MFCD31568537
  • Molecular Weight : 177.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl H2O
Storage temp. RT
Physical Form liquid
Shipping Temperature RT
Country of Origin RU
Product Link https://www.hit2lead.com/building-blocks/4091207

Safety Information

MSDS https://www.hit2lead.com/sds/4091207_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart