CH4H11AB8A96 ChemBridge Corporation

(1-azabicyclo[2.2.2]oct-2-ylmethyl)amine dihydrochloride

  • MDL number MFCD28892100
  • Cas Number : 860502-92-7
  • Molecular Weight : 213.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin RU
Product Link https://www.hit2lead.com/building-blocks/4038924

Safety Information

MSDS https://www.hit2lead.com/sds/4038924_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart