CH4H252D39F1 ChemBridge Corporation

3-propyl-1H-1,2,4-triazol-5-amine

  • MDL number MFCD07366086
  • Cas Number : 60016-62-8
  • Molecular Weight : 126.1597

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6741358

Safety Information

MSDS https://www.hit2lead.com/sds/6741358_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart