CH4H252D3A04 ChemBridge Corporation

4-methylpyrazolo[1,5-a]pyridine-3-carboxylic acid

  • MDL number MFCD12405114
  • Cas Number : 143803-93-4
  • Molecular Weight : 176.172

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4049566

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4049566
Availability details for
Quantity
  • Availability
  • Add to Cart