CH4H252D3A25 ChemBridge Corporation

1-oxa-9-azaspiro[5.5]undecane hydrochloride

  • MDL number MFCD16556302
  • Cas Number : 1414958-73-8
  • Molecular Weight : 191.6983

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4083046

Safety Information

MSDS https://www.hit2lead.com/sds/4083046_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart