CH4H30A32DC5 ChemBridge Corporation

N-methyl-1-(2-pyrrolidinyl)methanamine dihydrochloride

  • MDL number MFCD31440034
  • Molecular Weight : 187.1106

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4111536

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4111536
Availability details for
Quantity
  • Availability
  • Add to Cart