CH4H324AC5C7 ChemBridge Corporation

ethyl (6-chloro-2-pyridinyl)acetate

  • MDL number MFCD21604163
  • Cas Number : 174666-22-9
  • Molecular Weight : 199.6342

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. -20
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4110389

Safety Information

MSDS https://www.hit2lead.com/sds/4110389_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart