CH4H5F6D88D6 ChemBridge Corporation

2-(4-fluorophenyl)-5-methyl-1H-imidazole-4-carboxylic acid

  • MDL number MFCD11053198
  • Cas Number : 933751-61-2
  • Molecular Weight : 220.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin RU
Product Link https://www.hit2lead.com/building-blocks/4000036

Safety Information

MSDS https://www.hit2lead.com/sds/4000036_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart