CH4H5F6D88E0 ChemBridge Corporation

(3-exo)-8-azabicyclo[3.2.1]oct-3-ylmethanol hydrochloride

  • MDL number MFCD23381619
  • Cas Number : 1389264-20-3
  • Molecular Weight : 177.6717

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4046098

Safety Information

MSDS https://www.hit2lead.com/sds/4046098_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart