CH4H5F6D88F9 ChemBridge Corporation

methyl 6-amino-2-chloro-4-pyrimidinecarboxylate

  • MDL number MFCD14582103
  • Cas Number : 944129-00-4
  • Molecular Weight : 187.5837

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4110421

Safety Information

MSDS https://www.hit2lead.com/sds/4110421_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart