CH4H5F6D8912 ChemBridge Corporation

1-(4-piperidinyl)-3-azetidinol

  • MDL number MFCD16659802
  • Cas Number : 1147423-04-8
  • Molecular Weight : 156.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin RU
Product Link https://www.hit2lead.com/building-blocks/4110540

Safety Information

MSDS https://www.hit2lead.com/sds/4110540_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart