CH4H8C2C8593 ChemBridge Corporation

4,5,6,7-tetrahydropyrazolo[1,5-a]pyrazin-2-ylmethanol dihydrochloride

  • Molecular Weight : 226.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin RU
Product Link https://www.hit2lead.com/building-blocks/4081231

Safety Information

MSDS https://www.hit2lead.com/sds/4081231_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart