CH4H8C2C8599 ChemBridge Corporation

(1R,2S,9S)-7,11-diazatricyclo[7.3.1.0~2,7~]tridecane dihydrochloride

  • MDL number MFCD28891672
  • Molecular Weight : 253.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin RU
Product Link https://www.hit2lead.com/building-blocks/4036499

Safety Information

MSDS https://www.hit2lead.com/sds/4036499_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart