CH4H8C2C859E ChemBridge Corporation

1,3-dihydro-2-benzofuran-4-carboxylic acid

  • MDL number MFCD20642221
  • Molecular Weight : 164.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin RU
Product Link https://www.hit2lead.com/building-blocks/4074077

Safety Information

MSDS https://www.hit2lead.com/sds/4074077_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart