CH4H8C2C85A2 ChemBridge Corporation

(1,3-dimethyl-1H-pyrazol-4-yl)acetic acid

  • MDL number MFCD15143077
  • Cas Number : 1177342-49-2
  • Molecular Weight : 154.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin RU
Product Link https://www.hit2lead.com/building-blocks/4040478

Safety Information

MSDS https://www.hit2lead.com/sds/4040478_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart