CH4H8C2C85B5 ChemBridge Corporation

1-methyl-5-[(methylamino)methyl]pyrrolidin-2-one

  • MDL number MFCD18816866
  • Cas Number : 479065-35-5
  • Molecular Weight : 142.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Physical Form liquid
Shipping Temperature RT
Country of Origin RU
Product Link https://www.hit2lead.com/building-blocks/4111540

Safety Information

MSDS https://www.hit2lead.com/sds/4111540_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart