C12H316CE2AA Chemcia Scientific, LLC

4-Chloro-2-methoxy-benzoic acid methyl ester

  • MDL number MFCD09800922
  • Cas Number : 78955-90-5
  • Molecular Weight : 200.62328

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Pale Yellow Cystalline Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=9

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=9
Availability details for
Quantity
  • Availability
  • Add to Cart