C12H316CE2AB Chemcia Scientific, LLC

5-Bromo-4-chloro-2-fluoro-benzoic acid

  • MDL number MFCD00672928
  • Cas Number : 289038-22-8
  • Molecular Weight : 253.45616

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Pale Yellow Powder
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=2499

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=2499
Availability details for
Quantity
  • Availability
  • Add to Cart