C12H316CE2AC Chemcia Scientific, LLC

5-Bromo-4-chloro-2-hydroxy-benzoic acid

  • MDL number MFCD09475899
  • Cas Number : 142167-38-2
  • Molecular Weight : 251.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-bromo-4-chloro-2-hydroxybenzoic acid
Inchi Code 1S/C7H4BrClO3/c8-4-1-3(7(11)12)6(10)2-5(4)9/h1-2,10H,(H,11,12)
Inchi Key XUKLAPBPFXGIOW-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=13

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=13
Availability details for
Quantity
  • Availability
  • Add to Cart