C12H316CE2AD Chemcia Scientific, LLC

5-Bromo-4-chloro-2-methoxy-benzoic acid methyl ester

  • MDL number MFCD09800923
  • Cas Number : 951885-11-3
  • Molecular Weight : 279.51931

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form White Powder
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=14

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=14
Availability details for
Quantity
  • Availability
  • Add to Cart