C12H316CE2AE Chemcia Scientific, LLC

4-Chloro-2-methoxy-5-(4,4,5,5-tetramethyl-[1,3,2]dioxaborolan-2-yl)-benzoic acid methyl ester

  • MDL number MFCD19441823
  • Cas Number : 1070892-90-8
  • Molecular Weight : 326.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 4-chloro-2-methoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate
Inchi Code 1S/C15H20BClO5/c1-14(2)15(3,4)22-16(21-14)10-7-9(13(18)20-6)12(19-5)8-11(10)17/h7-8H,1-6H3
Inchi Key JWAUBYUEFQVXJK-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form White Crystalline Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=15

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=15
Availability details for
Quantity
  • Availability
  • Add to Cart