C12H316CE2B1 Chemcia Scientific, LLC

5-Bromo-4-Chloro-2-fluoro-benzoic acid methyl ester

  • MDL number MFCD09800902
  • Cas Number : 951884-02-9
  • Molecular Weight : 267.48325

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Pale Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=17

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=17
Availability details for
Quantity
  • Availability
  • Add to Cart