C12H316CE2B2 Chemcia Scientific, LLC

7-Methoxyindoline hydrochloride

  • MDL number MFCD11506101
  • Cas Number : 4770-43-8
  • Molecular Weight : 185.65509

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Salt Data HCl
Storage temp. 0~5
Physical Form Off-White Crystalline Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=18

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=18
Availability details for
Quantity
  • Availability
  • Add to Cart