C12H316CE2B3 Chemcia Scientific, LLC

2,3-Difluoro-6-methoxy-phenylamine hydrochloride

  • MDL number MFCD19441987
  • Cas Number : 1332581-65-3
  • Molecular Weight : 195.59771

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=42

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=42
Availability details for
Quantity
  • Availability
  • Add to Cart