C12H316CE2B4 Chemcia Scientific, LLC

2-Chloro-6-methoxy-phenylamine hydrochloride

  • MDL number MFCD19441992
  • Cas Number : 1332589-57-7
  • Molecular Weight : 194.06188

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Salt Data HCl
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=43

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=43
Availability details for
Quantity
  • Availability
  • Add to Cart