C12H316CE2B8 Chemcia Scientific, LLC

1-Chloro-3-methoxy-2-nitro-benzene

  • MDL number MFCD14455916
  • Cas Number : 5472-99-1
  • Molecular Weight : 187.58377

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=45

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=45
Availability details for
Quantity
  • Availability
  • Add to Cart