C12H316CE2BB Chemcia Scientific, LLC

2-Fluoro-6-methoxy-benzamide

  • MDL number MFCD00277474
  • Cas Number : 529512-81-0
  • Molecular Weight : 169.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-fluoro-6-methoxybenzamide
Inchi Code 1S/C8H8FNO2/c1-12-6-4-2-3-5(9)7(6)8(10)11/h2-4H,1H3,(H2,10,11)
Inchi Key BDJNFURQDZEPLK-UHFFFAOYSA-N

Description

Purity 98%
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=712

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=712
Availability details for
Quantity
  • Availability
  • Add to Cart