C12H316CE2BD Chemcia Scientific, LLC

N-(2-Fluoro-6-methoxyphenyl)formamide

  • MDL number MFCD19441833
  • Cas Number : 1313712-60-5
  • Molecular Weight : 169.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-fluoro-6-methoxyphenylformamide
Inchi Code 1S/C8H8FNO2/c1-12-7-4-2-3-6(9)8(7)10-5-11/h2-5H,1H3,(H,10,11)
Inchi Key KNFFYACFYGLVOV-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=47

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=47
Availability details for
Quantity
  • Availability
  • Add to Cart