C12H316CE2BE Chemcia Scientific, LLC

(S)-1-tert-Butoxycarbonylmethyl-4-oxo-piperidine-2-carboxylic acid

  • MDL number MFCD20921530
  • Cas Number : 1352721-92-6
  • Molecular Weight : 257.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2S)-1-(2-tert-butoxy-2-oxoethyl)-4-oxo-2-piperidinecarboxylic acid
Inchi Code 1S/C12H19NO5/c1-12(2,3)18-10(15)7-13-5-4-8(14)6-9(13)11(16)17/h9H,4-7H2,1-3H3,(H,16,17)/t9-/m0/s1
Inchi Key OZAWLTYEPXIRGI-VIFPVBQESA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=805

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=805
Availability details for
Quantity
  • Availability
  • Add to Cart