C12H316CE2BF Chemcia Scientific, LLC

Bis-(4-chloro-phenyl)-diazene

  • MDL number MFCD00018598
  • Cas Number : 1602-00-2
  • Molecular Weight : 251.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1,2-bis(4-chlorophenyl)diazene
Inchi Code 1S/C12H8Cl2N2/c13-9-1-5-11(6-2-9)15-16-12-7-3-10(14)4-8-12/h1-8H/b16-15+
Inchi Key XHQLXCFUPJSGOE-FOCLMDBBSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Red Crystalline Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=125

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=125
Availability details for
Quantity
  • Availability
  • Add to Cart