C12H316CE2C5 Chemcia Scientific, LLC

4-Amino-5,6-dihydro-2H-pyridine-1,3-dicarboxylic acid 1-tert-butyl ester 3-ethyl ester

  • MDL number MFCD18434501
  • Cas Number : 947403-75-0
  • Molecular Weight : 270.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-tert-butyl 3-ethyl 4-amino-5,6-dihydro-1,3(2H)-pyridinedicarboxylate
Inchi Code 1S/C13H22N2O4/c1-5-18-11(16)9-8-15(7-6-10(9)14)12(17)19-13(2,3)4/h5-8,14H2,1-4H3
Inchi Key GJAQACJJJRXXED-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=136

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=136
Availability details for
Quantity
  • Availability
  • Add to Cart