C12H316CE2C8 Chemcia Scientific, LLC

2-Amino-cyclopent-1-enecarboxylic acid methyl ester

  • MDL number MFCD19204033
  • Cas Number : 52909-60-1
  • Molecular Weight : 141.17122

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Pale Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=145

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=145
Availability details for
Quantity
  • Availability
  • Add to Cart