C12H316CE2CA Chemcia Scientific, LLC

3-(4-Methoxy-benzyl)-5,6,7,8-tetrahydro-1H-quinazoline-2,4-dione

  • MDL number MFCD19441859
  • Cas Number : 1313712-33-2
  • Molecular Weight : 286.33346

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Off-White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=148

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=148
Availability details for
Quantity
  • Availability
  • Add to Cart