C12H316CE2D2 Chemcia Scientific, LLC

2-Methyl-5,6,7,8-tetrahydro-3H-quinazolin-4-one

  • MDL number MFCD01685799
  • Cas Number : 19178-21-3
  • Molecular Weight : 164.20879

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Off-White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=153

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=153
Availability details for
Quantity
  • Availability
  • Add to Cart