C12H316CE2D5 Chemcia Scientific, LLC

2-Trifluoromethyl-5,6,7,8-tetrahydro-3H-quinazolin-4-one

  • MDL number MFCD18909471
  • Cas Number : 147750-20-7
  • Molecular Weight : 218.18008

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. 0~5
Physical Form Off-White Crystalline Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=91

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=91
Availability details for
Quantity
  • Availability
  • Add to Cart