C12H316CE2DA Chemcia Scientific, LLC

5-Bromo-3-(2,2,2-trifluoro-acetylamino)-thiophene-2-carboxylic acid methyl ester

  • MDL number MFCD19441860
  • Cas Number : 852330-31-5
  • Molecular Weight : 332.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 5-bromo-3-[(trifluoroacetyl)amino]-2-thiophenecarboxylate
Inchi Code 1S/C8H5BrF3NO3S/c1-16-6(14)5-3(2-4(9)17-5)13-7(15)8(10,11)12/h2H,1H3,(H,13,15)
Inchi Key AUIUYLZMPYTXPA-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=158

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=158
Availability details for
Quantity
  • Availability
  • Add to Cart