C12H316CE2DD Chemcia Scientific, LLC

2-Methyl-6,7-dihydro-5H-cyclopentapyrimidin-4-ol

  • MDL number MFCD16872922
  • Cas Number : 65818-01-1
  • Molecular Weight : 150.1817

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=668

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=668
Availability details for
Quantity
  • Availability
  • Add to Cart