C12H316CE2E3 Chemcia Scientific, LLC

3-(4-Methoxy-benzyl)-1H-thieno[3,2-d]pyrimidine-2,4-dione

  • MDL number MFCD14703386
  • Cas Number : 1223888-84-3
  • Molecular Weight : 288.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-(4-methoxybenzyl)thieno[3,2-d]pyrimidine-2,4(1H,3H)-dione
Inchi Code 1S/C14H12N2O3S/c1-19-10-4-2-9(3-5-10)8-16-13(17)12-11(6-7-20-12)15-14(16)18/h2-7H,8H2,1H3,(H,15,18)
Inchi Key GBVIVVHEVHMWEO-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=163

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=163
Availability details for
Quantity
  • Availability
  • Add to Cart